Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081033.1 | NP_001074502.1 | 11 - 12 | 1983 | 129 | |
GenBank mRNA | AK042383.1 | - | 8 - 9 | 761 | 129 | |
BC158125.1 | - | 11 - 12 | 2123 | 9088197 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |