Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178693.4 | NP_848808.1 | 5 - 6 | 557 | 99 | |
GenBank mRNA | AK039422.1 | - | 5 - 6 | 558 | 99 | |
AK039676.1 | - | 5 - 6 | 558 | 99 | ||
AK043574.1 | - | 5 - 6 | 533 | 99 | ||
AK050459.1 | - | 5 - 6 | 567 | 99 | ||
BC139373.1 | - | 5 - 6 | 600 | 8862395 | 99 | |
BC139376.1 | - | 5 - 6 | 597 | 8862398 | 99 | |
BC145615.1 | - | 5 - 6 | 571 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |