Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3glb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289709.1 | NP_001276638.1 | 2 - 3 | 313 | 101 | |
NM_001289710.1 | NP_001276639.1 | 2 - 3 | 313 | 101 | ||
NM_139302.2 | NP_647463.1 | 2 - 3 | 313 | 101 | ||
GenBank mRNA | AF426314.1 | - | 2 - 3 | 210 | 101 | |
AK129455.1 | - | 2 - 3 | 241 | 101 | ||
AK152409.1 | - | 2 - 3 | 243 | 101 | ||
AK152502.1 | - | 2 - 3 | 263 | 102 | ||
AK155248.1 | - | 2 - 3 | 238 | 101 | ||
AK167762.1 | - | 2 - 3 | 286 | 101 | ||
BC024477.1 | - | 2 - 3 | 228 | 4977001 | 101 | |
BC028859.1 | - | 2 - 3 | 302 | 4167974 | 101 | |
CX216029.1 | - | 2 - 3 | 303 | 101 | ||
EF676083.1 | - | 2 - 3 | 289 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |