Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trub2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290496.1 | NP_001277425.1 | 3 - 4 | 370 | 72 | |
NM_145520.5 | NP_663495.3 | 3 - 4 | 370 | 72 | ||
GenBank mRNA | AK087102.1 | - | 3 - 4 | 352 | 72 | |
AK090004.1 | - | 1202 | 72 | |||
AK167294.1 | - | 3 - 4 | 346 | 72 | ||
BC015285.1 | - | 3 - 4 | 349 | 4239390 | 72 | |
BC024075.1 | - | 3 - 4 | 339 | 5097466 | 72 | |
BU053791.1 | - | 3 - 4 | 322 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |