Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285458.1 | NP_001272387.1 | 9 - 10 | 3459 | 111 | |
NM_001285459.1 | NP_001272388.1 | 9 - 10 | 3083 | 111 | ||
NM_010230.2 | NP_034360.2 | 11 - 12 | 3753 | 111 | ||
GenBank mRNA | BC138036.1 | - | 8 - 9 | 3365 | 8861055 | 111 |
BC171945.1 | - | 8 - 9 | 3359 | 9054231 | 111 | |
CK391697.1 | - | 5 - 6 | 385 | 111 | ||
CO814739.2 | - | 4 - 5 | 310 | 111 | ||
X53599.1 | - | 11 - 12 | 3713 | 111 | ||
X62379.1 | - | 9 - 10 | 3089 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |