Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153805.1 | NP_722500.1 | 5 - 6 | 646 | 77 | |
GenBank mRNA | AK142182.1 | - | 4 - 5 | 1162 | 77 | |
AK142988.1 | - | 4 - 5 | 1218 | 77 | ||
AK154801.1 | - | 4 - 5 | 1183 | 77 | ||
BC034126.1 | - | 5 - 6 | 646 | 4912536 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |