Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012310.2 | NP_001012310.2 | 1 - 2 | 85 | 65 | |
GenBank mRNA | AK149369.1 | - | 1 - 2 | 91 | 65 | |
BC035527.1 | - | 1 - 2 | 86 | 65 | ||
BC055027.1 | - | 1 - 2 | 78 | 65 | ||
BC069939.1 | - | 1 - 2 | 74 | 5053384 | 65 | |
BC089616.1 | - | 1 - 2 | 85 | 6760348 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |