Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244891.1 | NP_001231820.1 | 10 - 11 | 1199 | 88 | |
NM_001244903.1 | NP_001231832.1 | 9 - 10 | 1146 | 88 | ||
NM_017368.3 | NP_059064.2 | 10 - 11 | 1298 | 88 | ||
NM_198683.2 | NP_941955.1 | 9 - 10 | 1146 | 88 | ||
GenBank mRNA | AJ007987.1 | - | 8 - 9 | 891 | 88 | |
AK143698.1 | - | 10 - 11 | 1100 | 88 | ||
AK156725.1 | - | 9 - 10 | 1059 | 88 | ||
BC021393.1 | - | 9 - 10 | 1011 | 88 | ||
X61451.1 | - | 5 - 6 | 594 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |