Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080944.3 | NP_001074413.2 | 11 - 12 | 943 | 114 | |
GenBank mRNA | AK040903.1 | - | 12 - 13 | 1112 | 114 | |
AK086607.1 | - | 11 - 12 | 977 | 114 | ||
AK155448.1 | - | 11 - 12 | 953 | 114 | ||
AK169967.1 | - | 11 - 12 | 966 | 114 | ||
AK173307.1 | - | 11 - 12 | 943 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter Mus musculus Probe spans exons |