Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080944.3 | NP_001074413.2 | 2 - 3 | 145 | 74 | |
GenBank mRNA | AK040903.1 | - | 3 - 4 | 314 | 74 | |
AK086607.1 | - | 2 - 3 | 178 | 74 | ||
AK155448.1 | - | 2 - 3 | 156 | 74 | ||
AK169967.1 | - | 2 - 3 | 168 | 74 | ||
AK173307.1 | - | 2 - 3 | 145 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Mus musculus Probe spans exons |