Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199141.1 | NP_001186070.1 | 8 - 9 | 1030 | 94 | |
NM_144516.3 | NP_653099.2 | 7 - 8 | 868 | 94 | ||
GenBank mRNA | AK028931.1 | - | 5 - 6 | 452 | 94 | |
AK077732.1 | - | 7 - 8 | 715 | 94 | ||
AK145892.1 | - | 7 - 8 | 662 | 94 | ||
BC058999.1 | - | 8 - 9 | 996 | 6827880 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |