Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.1 ST Array |
10476668 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0200016884.mm.1 |
NM_001164813.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016884.mm.1 |
NM_001164813.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016884.mm.1 |
NM_001166640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016884.mm.1 |
NM_001166640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016884.mm.1 |
NM_181417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016884.mm.1 |
NM_181417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016884.mm.1 |
TC0200002146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016887.mm.1 |
NM_001164813.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016887.mm.1 |
NM_001164813.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016887.mm.1 |
NM_001166640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016887.mm.1 |
NM_001166640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016887.mm.1 |
NM_181417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016887.mm.1 |
NM_181417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016887.mm.1 |
TC0200002146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016888.mm.1 |
NM_001166640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016888.mm.1 |
NM_001166640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016888.mm.1 |
NM_181417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016888.mm.1 |
NM_181417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016888.mm.1 |
TC0200002146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016890.mm.1 |
NM_181417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016890.mm.1 |
NM_181417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016890.mm.1 |
TC0200002146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016882.mm.1 |
NM_001164813.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016882.mm.1 |
NM_001164813.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016882.mm.1 |
TC0200002146.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016885.mm.1 |
NM_001164813.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016885.mm.1 |
NM_001164813.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016885.mm.1 |
NM_001166640.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016885.mm.1 |
NM_001166640.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016885.mm.1 |
NM_181417.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016885.mm.1 |
NM_181417.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016885.mm.1 |
TC0200002146.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1442405_PM_at |
- |
- |
Mouse Expression Set 430 |
1442405_at |
- |
- |