Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200038015.mm.1 |
NM_080459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038015.mm.1 |
NM_080459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038015.mm.1 |
TC0200004486.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200038018.mm.1 |
NM_080459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038018.mm.1 |
NM_080459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038018.mm.1 |
TC0200004486.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200038019.mm.1 |
NM_080459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038019.mm.1 |
NM_080459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038019.mm.1 |
TC0200004486.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200038016.mm.1 |
NM_080459.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038016.mm.1 |
NM_080459.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200038016.mm.1 |
TC0200004486.mm.1 |
MTA TC |