Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163701.1 | NP_001157173.1 | 17 - 18 | 2665 | 82 | |
NM_177294.5 | NP_796268.3 | 18 - 19 | 2742 | 82 | ||
GenBank mRNA | AK122505.1 | - | 18 - 19 | 2709 | 82 | |
AK154007.1 | - | 17 - 18 | 2649 | 82 | ||
AK161487.1 | - | 18 - 19 | 2725 | 82 | ||
AK164658.1 | - | 18 - 19 | 2726 | 82 | ||
AK166232.1 | - | 17 - 18 | 2648 | 82 | ||
AK167589.1 | - | 17 - 18 | 2648 | 82 | ||
AK171233.1 | - | 17 - 18 | 2621 | 82 | ||
BC051680.1 | - | 4 - 5 | 658 | 82 | ||
BC088899.1 | - | 16 - 17 | 2470 | 82 | ||
BC117973.1 | - | 17 - 18 | 2634 | 40098643 | 82 | |
BC117974.1 | - | 17 - 18 | 2634 | 40098644 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |