Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap4e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175550.3 | NP_780759.2 | 14 - 15 | 1890 | 69 | |
GenBank mRNA | AK036944.1 | - | 14 - 15 | 1871 | 69 | |
AK144636.1 | - | 14 - 15 | 1871 | 69 | ||
BC042530.1 | - | 1 - 2 | 81 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |