Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177244.3 | NP_796218.2 | 11 - 12 | 2291 | 93 | |
GenBank mRNA | AK220423.1 | - | 7 - 8 | 1139 | 93 | |
BC023501.1 | - | 4 - 5 | 424 | 93 | ||
BC034292.1 | - | 8 - 9 | 1398 | 93 | ||
BC069943.1 | - | 10 - 11 | 1784 | 93 | ||
BC075689.1 | - | 11 - 12 | 2291 | 30609755 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |