Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp46a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010010.1 | NP_034140.1 | 4 - 5 | 428 | 66 | |
GenBank mRNA | AF094479.1 | - | 4 - 5 | 428 | 66 | |
AK140740.1 | - | 4 - 5 | 433 | 66 | ||
BC018307.1 | - | 4 - 5 | 451 | 4195579 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |