Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yme1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013771.5 | NP_038799.1 | 2 - 3 | 359 | 154 | |
GenBank mRNA | AF090430.1 | - | 2 - 3 | 337 | 154 | |
AK032273.1 | - | 2 - 3 | 347 | 154 | ||
AK045141.1 | - | 2 - 3 | 344 | 154 | ||
AK079175.1 | - | 2 - 3 | 319 | 154 | ||
AK137665.1 | - | 2 - 3 | 311 | 154 | ||
AY136286.1 | - | 2 - 3 | 175 | 154 | ||
AY136287.1 | - | 2 - 3 | 328 | 154 | ||
BB624412.1 | - | 2 - 3 | 359 | 154 | ||
BC007128.1 | - | 2 - 3 | 320 | 3591123 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |