Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yme1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013771.5 | NP_038799.1 | 6 - 7 | 879 | 68 | |
GenBank mRNA | AF090430.1 | - | 6 - 7 | 857 | 68 | |
AK032273.1 | - | 6 - 7 | 867 | 68 | ||
AK079175.1 | - | 6 - 7 | 839 | 68 | ||
AK137665.1 | - | 6 - 7 | 831 | 68 | ||
AY136286.1 | - | 6 - 7 | 695 | 68 | ||
AY136287.1 | - | 6 - 7 | 848 | 68 | ||
BC007128.1 | - | 6 - 7 | 840 | 3591123 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |