Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glp2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175681.3 | NP_783612.2 | 2 - 3 | 440 | 91 | |
GenBank mRNA | AY255567.1 | - | 1 - 2 | 54 | 91 | |
AY605231.1 | - | 2 - 3 | 159 | 91 | ||
BC044746.2 | - | 2 - 3 | 440 | 5363416 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |