Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1976282 |
AK144419.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1976282 |
BU559819.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1976282 |
NM_011102.3 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
827794 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
928342 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
773887 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
634675 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
403069 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
860917 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
342220 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
617447 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
162537 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906716 |
AB073619.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906716 |
AK003653.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906716 |
BC006879.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906716 |
NM_001110271.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906716 |
NM_001110272.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
906716 |
NM_145919.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
63795 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
668277 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
409104 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
243973 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
3595 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
550589 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
67058 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
971641 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
66877 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
926908 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
543228 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
286841 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
697685 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
6430 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
44826 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
80251 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
70954 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
26728 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
118943 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
135242 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1976282 |
AK144419.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1976282 |
BU559819.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1976282 |
NM_011102.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P290290 |
L28035.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P290290 |
X67129.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P290290 |
AK144419.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P290290 |
NM_011102.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P290290 |
L28035.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P290290 |
X67129.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P290290 |
AK144419.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P290290 |
NM_011102.3 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P126414 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984472 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984479 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984428 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982670 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982661 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984486 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984422 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982685 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984446 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984481 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984471 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982664 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982677 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982665 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984436 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984484 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984467 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984478 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982675 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984440 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984488 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984443 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982668 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984424 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984430 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984474 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984437 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984473 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984465 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984482 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984470 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984480 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984448 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982680 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984427 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984452 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982673 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984477 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984483 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984420 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982662 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984468 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982660 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984451 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984423 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982679 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984476 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984435 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984429 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984441 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984489 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984469 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32982678 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984426 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P32984444 |
- |
- |