Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172584.3 | NP_766172.1 | 4 - 5 | 417 | 94 | |
GenBank mRNA | AK038913.1 | - | 4 - 5 | 418 | 94 | |
BC056464.1 | - | 4 - 5 | 398 | 5707377 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |