Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042630.2 | NP_001036095.1 | 1 - 2 | 138 | 66 | |
NM_001276257.1 | NP_001263186.1 | 1 - 2 | 138 | 66 | ||
NM_007947.3 | NP_031973.2 | 1 - 2 | 138 | 66 | ||
GenBank mRNA | AK021014.1 | - | 1 - 2 | 136 | 66 | |
AK136629.1 | - | 1 - 2 | 141 | 66 | ||
AK162355.1 | - | 1 - 2 | 137 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |