Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200026393.mm.1 |
NM_133835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026393.mm.1 |
NM_133835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026393.mm.1 |
TC0200003174.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026397.mm.1 |
NM_133835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026397.mm.1 |
NM_133835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026397.mm.1 |
TC0200003174.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026395.mm.1 |
NM_133835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026395.mm.1 |
NM_133835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026395.mm.1 |
TC0200003174.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026399.mm.1 |
NM_133835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026399.mm.1 |
NM_133835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026399.mm.1 |
TC0200003174.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026398.mm.1 |
NM_133835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026398.mm.1 |
NM_133835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026398.mm.1 |
TC0200003174.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026396.mm.1 |
NM_133835.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026396.mm.1 |
NM_133835.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026396.mm.1 |
TC0200003174.mm.1 |
MTA TC |