Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199146.1 | NP_001186075.1 | 9 - 10 | 1321 | 70 | |
NM_001199147.1 | NP_001186076.1 | 9 - 10 | 1390 | 70 | ||
NM_153410.5 | NP_700459.2 | 9 - 10 | 1321 | 70 | ||
GenBank mRNA | AK048691.1 | - | 9 - 10 | 1315 | 70 | |
AK082985.1 | - | 9 - 10 | 1265 | 70 | ||
AK084631.1 | - | 9 - 10 | 1263 | 70 | ||
BC027004.1 | - | 6 - 7 | 672 | 70 | ||
BC071197.1 | - | 9 - 10 | 1387 | 6465025 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |