Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026184.2 | NP_080460.2 | 5 - 6 | 625 | 63 | |
GenBank mRNA | AF489856.1 | - | 5 - 6 | 431 | 63 | |
AK004981.1 | - | 602 | 63 | |||
AK006886.1 | - | 5 - 6 | 488 | 63 | ||
AK041951.1 | - | 5 - 6 | 601 | 63 | ||
AK160664.1 | - | 5 - 6 | 641 | 63 | ||
BC058721.1 | - | 5 - 6 | 625 | 5705507 | 63 | |
BC111900.1 | - | 5 - 6 | 595 | 40062137 | 63 | |
BC112426.1 | - | 5 - 6 | 595 | 40062136 | 63 | |
BC145888.1 | - | 5 - 6 | 576 | 40131191 | 63 | |
BC145890.1 | - | 5 - 6 | 576 | 40131193 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |