Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrdl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114385.1 | NP_001107857.1 | 9 - 10 | 1478 | 92 | |
GenBank mRNA | AF321853.1 | - | 8 - 9 | 954 | 92 | |
BC050818.1 | - | 8 - 9 | 968 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |