Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080848.2 | NP_543124.1 | 6 - 7 | 659 | 80 | |
GenBank mRNA | AF416510.1 | - | 6 - 7 | 664 | 80 | |
AK075937.1 | - | 6 - 7 | 602 | 80 | ||
AK144091.1 | - | 6 - 7 | 603 | 80 | ||
BC016103.1 | - | 6 - 7 | 594 | 4919781 | 80 | |
BC025801.1 | - | 6 - 7 | 636 | 5132525 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |