Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nqo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163241.1 | NP_001156713.1 | 2 - 3 | 233 | 113 | |
NM_001163242.1 | NP_001156714.1 | 2 - 3 | 162 | 113 | ||
NM_020282.3 | NP_064678.1 | 3 - 4 | 315 | 113 | ||
GenBank mRNA | AK010842.1 | - | 2 - 3 | 205 | 113 | |
AK049781.1 | - | 2 - 3 | 171 | 113 | ||
BC027629.1 | - | 3 - 4 | 223 | 1225122 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |