Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010726.2 | NP_034856.1 | 7 - 8 | 906 | 66 | |
GenBank mRNA | AF023463.1 | - | 7 - 8 | 871 | 66 | |
AK146753.1 | - | 7 - 8 | 891 | 66 | ||
AK151372.1 | - | 7 - 8 | 854 | 66 | ||
AK164495.1 | - | 7 - 8 | 857 | 66 | ||
AK165412.1 | - | 7 - 8 | 850 | 66 | ||
AK169213.1 | - | 7 - 8 | 848 | 66 | ||
BC002018.1 | - | 7 - 8 | 883 | 3489939 | 66 | |
D88670.1 | - | 7 - 8 | 866 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |