Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dab2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001602.2 | NP_001001602.2 | 5 - 6 | 1365 | 86 | |
NM_001114124.2 | NP_001107596.1 | 7 - 8 | 1489 | 86 | ||
NM_001114125.1 | NP_001107597.1 | 7 - 8 | 1341 | 86 | ||
NM_001290637.1 | NP_001277566.1 | 7 - 8 | 1396 | 86 | ||
NM_001290639.1 | NP_001277568.1 | 5 - 6 | 1024 | 86 | ||
NM_001290640.1 | NP_001277569.1 | 5 - 6 | 1329 | 86 | ||
NM_001290641.1 | NP_001277570.1 | 7 - 8 | 1622 | 86 | ||
GenBank mRNA | AK122548.1 | - | 2 - 3 | 329 | 86 | |
AK147464.1 | - | 7 - 8 | 1421 | 86 | ||
AK147593.1 | - | 7 - 8 | 1623 | 86 | ||
AK155296.1 | - | 7 - 8 | 1397 | 86 | ||
AY305656.1 | - | 5 - 6 | 1082 | 86 | ||
AY305657.1 | - | 5 - 6 | 1365 | 86 | ||
AY305658.1 | - | 5 - 6 | 1024 | 86 | ||
BC094373.1 | - | 2 - 3 | 186 | 86 | ||
BC118530.1 | - | 5 - 6 | 990 | 40098202 | 86 | |
DQ473307.1 | - | 7 - 8 | 1226 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |