Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177268.4 | NP_796242.2 | 4 - 5 | 1528 | 90 | |
GenBank mRNA | BC021802.1 | - | 4 - 5 | 657 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |