Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3glb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289710.1 | NP_001276639.1 | 6 - 7 | 696 | 54 | |
GenBank mRNA | AK152502.1 | - | 6 - 7 | 647 | 54 | |
BC028859.1 | - | 6 - 7 | 685 | 4167974 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |