Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164791.1 | NP_001158263.1 | 8 - 9 | 863 | 81 | |
NM_023057.5 | NP_075544.1 | 8 - 9 | 927 | 81 | ||
NM_178084.4 | NP_835185.2 | 8 - 9 | 863 | 81 | ||
GenBank mRNA | AB049731.1 | - | 7 - 8 | 710 | 81 | |
AB049732.1 | - | 7 - 8 | 710 | 81 | ||
AK045444.1 | - | 8 - 9 | 788 | 81 | ||
AK087363.1 | - | 8 - 9 | 803 | 81 | ||
AK132186.1 | - | 8 - 9 | 928 | 81 | ||
AK162819.1 | - | 8 - 9 | 728 | 81 | ||
BC023718.1 | - | 8 - 9 | 808 | 5346593 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |