Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198872.1 | NP_001185801.1 | 12 - 13 | 1191 | 114 | |
NM_001198873.1 | NP_001185802.1 | 12 - 13 | 1176 | 114 | ||
NM_001198874.1 | NP_001185803.1 | 10 - 11 | 1122 | 114 | ||
NM_001198875.1 | NP_001185804.1 | 10 - 11 | 1080 | 114 | ||
NM_001198876.1 | NP_001185805.1 | 9 - 10 | 1062 | 114 | ||
NM_001198877.1 | NP_001185806.1 | 12 - 13 | 1180 | 114 | ||
NM_001198878.1 | NP_001185807.1 | 12 - 13 | 1165 | 114 | ||
NM_010064.4 | NP_034194.1 | 9 - 10 | 1051 | 114 | ||
GenBank mRNA | AF063231.1 | - | 9 - 10 | 990 | 114 | |
AK088015.1 | - | 9 - 10 | 1022 | 114 | ||
AK141837.1 | - | 10 - 11 | 1090 | 114 | ||
AK150173.1 | - | 9 - 10 | 1012 | 114 | ||
AK161831.1 | - | 5 - 6 | 1712 | 114 | ||
AK164322.1 | - | 10 - 11 | 1046 | 114 | ||
AK164437.1 | - | 9 - 10 | 986 | 114 | ||
AK168730.1 | - | 9 - 10 | 1026 | 114 | ||
BC139374.1 | - | 9 - 10 | 946 | 8862396 | 114 | |
BC139375.1 | - | 9 - 10 | 946 | 8862397 | 114 | |
GU992208.1 | - | 12 - 13 | 1082 | 114 | ||
GU992209.1 | - | 12 - 13 | 1067 | 114 | ||
GU992210.1 | - | 10 - 11 | 971 | 114 | ||
GU992211.1 | - | 12 - 13 | 1120 | 114 | ||
GU992212.1 | - | 12 - 13 | 1105 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |