Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200029961.mm.1 |
NM_001141982.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029961.mm.1 |
NM_001141982.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029961.mm.1 |
TC0200003526.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200029958.mm.1 |
NM_001141981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029958.mm.1 |
NM_001141981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029958.mm.1 |
NM_001141982.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029958.mm.1 |
NM_001141982.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029958.mm.1 |
NM_030243.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029958.mm.1 |
NM_030243.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200029958.mm.1 |
TC0200003526.mm.1 |
MTA TC |