Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cage1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027724.2 | NP_082000.2 | 2 - 3 | 371 | 96 | |
GenBank mRNA | AK016953.1 | - | 2 - 3 | 372 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |