Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpped2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143683.1 | NP_001137155.1 | 5 - 6 | 1131 | 76 | |
NM_029837.1 | NP_084113.1 | 5 - 6 | 844 | 76 | ||
GenBank mRNA | AK012553.1 | - | 5 - 6 | 845 | 76 | |
AK161666.1 | - | 5 - 6 | 1132 | 76 | ||
AK165186.1 | - | 5 - 6 | 1328 | 76 | ||
BC020182.1 | - | 4 - 5 | 859 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |