Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011576.1 | NP_035706.1 | 7 - 8 | 982 | 126 | |
GenBank mRNA | AF004833.1 | - | 6 - 7 | 751 | 126 | |
AK028356.1 | - | 7 - 8 | 982 | 126 | ||
AK167522.1 | - | 7 - 8 | 844 | 126 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |