Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029911.4 | NP_084187.2 | 1 - 2 | 162 | 109 | |
GenBank mRNA | AK036066.1 | - | 1 - 2 | 436 | 109 | |
AK082153.1 | - | 1 - 2 | 163 | 109 | ||
BC132487.1 | - | 1 - 2 | 104 | 40130764 | 109 | |
BC137869.1 | - | 1 - 2 | 104 | 8860888 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus 1700 Probe spans exons |