Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033213.1 | NP_001028385.1 | 15 - 16 | 1880 | 67 | |
GenBank mRNA | AK046740.1 | - | 15 - 16 | 1850 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |