Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181416.3 | NP_852081.2 | 11 - 12 | 2049 | 143 | |
GenBank mRNA | AK032639.1 | - | 4 - 5 | 460 | 143 | |
AK129034.1 | - | 2044 | 143 | |||
BC050875.1 | - | 11 - 12 | 2003 | 6511064 | 143 | |
BU052980.1 | - | 1 - 2 | 142 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |