Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cerkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048176.1 | NP_001041641.1 | 1 - 2 | 287 | 69 | |
GenBank mRNA | BY742285.1 | - | 1 - 2 | 297 | 69 | |
HQ426680.1 | - | 1 - 2 | 287 | 69 | ||
HQ426681.1 | - | 1 - 2 | 287 | 69 | ||
HQ426682.1 | - | 1 - 2 | 287 | 69 | ||
HQ426684.1 | - | 1 - 2 | 287 | 69 | ||
HQ426685.1 | - | 1 - 2 | 287 | 69 | ||
HQ426686.1 | - | 1 - 2 | 287 | 69 | ||
HQ426688.1 | - | 1 - 2 | 287 | 69 | ||
HQ426689.1 | - | 1 - 2 | 287 | 69 | ||
HQ426690.1 | - | 1 - 2 | 287 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |