Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080853.3 | NP_543129.3 | 1 - 2 | 889 | 119 | |
GenBank mRNA | AF324864.1 | - | 1 - 2 | 722 | 119 | |
AK043753.1 | - | 1 - 2 | 430 | 119 | ||
AK045409.1 | - | 1 - 2 | 652 | 119 | ||
BC038375.1 | - | 1 - 2 | 268 | 5357143 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |