Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081132.1 | NP_001074601.1 | 15 - 16 | 3137 | 148 | |
GenBank mRNA | BC039278.1 | - | 4 - 5 | 800 | 148 | |
BC138358.1 | - | 15 - 16 | 3100 | 8861378 | 148 | |
BC138359.1 | - | 15 - 16 | 3100 | 8861379 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |