Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calcrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018782.2 | NP_061252.2 | 7 - 8 | 816 | 86 | |
GenBank mRNA | AB015595.1 | - | 7 - 8 | 820 | 86 | |
AF146525.1 | - | 5 - 6 | 493 | 86 | ||
AF209905.1 | - | 3 - 4 | 312 | 86 | ||
AK137210.1 | - | 7 - 8 | 815 | 86 | ||
BC138537.1 | - | 4 - 5 | 443 | 8861557 | 86 | |
BC138540.1 | - | 4 - 5 | 443 | 8861560 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |