Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200033389.mm.1 |
NM_001177319.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033389.mm.1 |
NM_001177319.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033389.mm.1 |
NM_001177320.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033389.mm.1 |
NM_001177320.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033389.mm.1 |
NM_011576.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033389.mm.1 |
NM_011576.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033389.mm.1 |
TC0200003866.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200033395.mm.1 |
NM_011576.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033395.mm.1 |
NM_011576.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033395.mm.1 |
TC0200003866.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200033392.mm.1 |
NM_001177319.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033392.mm.1 |
NM_001177319.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033392.mm.1 |
NM_001177320.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033392.mm.1 |
NM_001177320.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033392.mm.1 |
NM_011576.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033392.mm.1 |
NM_011576.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200033392.mm.1 |
TC0200003866.mm.1 |
MTA TC |