Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160214.1 | NP_001153686.1 | 11 - 12 | 2655 | 168 | |
NM_211355.2 | NP_997594.1 | 11 - 12 | 2616 | 168 | ||
GenBank mRNA | AK051239.1 | - | 11 - 12 | 2541 | 168 | |
AK148868.1 | - | 4 - 5 | 488 | 168 | ||
AK156272.1 | - | 11 - 12 | 2578 | 168 | ||
AK173331.1 | - | 10 - 11 | 4324 | 168 | ||
BC044817.1 | - | 1 - 2 | 376 | 168 | ||
BC064465.1 | - | 11 - 12 | 2314 | 6825400 | 168 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |