Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_211355.2 | NP_997594.1 | 7 - 8 | 2059 | 92 | |
GenBank mRNA | AK051239.1 | - | 7 - 8 | 1984 | 92 | |
AK156272.1 | - | 7 - 8 | 2021 | 92 | ||
BC064465.1 | - | 7 - 8 | 1757 | 6825400 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |