Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008817.2 | NP_032843.2 | 8 - 9 | 853 | 72 | |
GenBank mRNA | AB003040.1 | - | 8 - 9 | 850 | 72 | |
AK044602.1 | - | 9 - 10 | 917 | 72 | ||
AK122238.1 | - | 8 - 9 | 832 | 72 | ||
AK141217.1 | - | 8 - 9 | 854 | 72 | ||
AK163845.1 | - | 8 - 9 | 856 | 72 | ||
BC048778.1 | - | 3 - 4 | 231 | 6308481 | 71 | |
BC072661.1 | - | 8 - 9 | 841 | 30543809 | 72 | |
BC085183.1 | - | 8 - 9 | 838 | 30663888 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |